α,α,α-Trifluoro-4-iodotoluene ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Iodobenzotrifluoride, 1-Iodo-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-iodotoluene
A12189.03 A12189.06 A12189.14
A12189.03EA 785 SEK
A12189.03 A12189.06 A12189.14
α,α,α-Trifluoro-4-iodotoluene ≥98%
α,α,α-Trifluoro-4-iodotoluene
Purity: 98% CAS No.: 455-13-0 Molecular Formula: C7H4F3I Formula Weight: 272.01 Flash Point: 68°(154°F) Refractive Index: 1.5180 Melting Point: -8° Boiling Point: 68-70°/15mm Beilstein Reference: 1944013 MDL No.: MFCD00039398
Formel: C₇H₄F₃I
Kokpunkt: 68…70 °C (15 mmHg)
Smältpunkt: –8 °C
Densitet: 1,851
Flampunkt: 68 °C (154 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00039398
CAS nummer: 455-13-0
EINECS: 207-234-4