3-Chloro-2-methylanisole ≥97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Chloro-6-methoxytoluene, 1-Chloro-3-methoxy-2-methylbenzene
B22203.06 B22203.14 B22203.22
B22203.06EA 1275 SEK
B22203.06 B22203.14 B22203.22
3-Chloro-2-methylanisole ≥97%
3-Chloro-2-methylanisole
Purity: 97% CAS No.: 3260-88-6 Molecular Formula: C8H9ClO Formula Weight: 156.61 Flash Point: 92°(197°F) Refractive Index: 1.5395 Melting Point: -3° Boiling Point: 212-215° Beilstein Reference: 1862220 UN No.: 1993 MDL No.: MFCD00070772
Formel: C₈H₉ClO
MW: 156.61 g/mol
Kokpunkt: 212…215 °C
Smältpunkt: –3 °C
Densitet: 1.160
Flampunkt: 92 °C (197 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00070772
CAS nummer: 3260-88-6
EINECS: 221-862-6
ADR: 3 III