2-Chloro-5-nitrobenzotrifluoride ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Chloro-5-nitrobenzotrifluoride, 2-Chloro-α,α,α-trifluoro-5-nitrotoluene, 1-Chloro-4-nitro-2-(trifluoromethyl)benzene
L06623.14 L06623.22
L06623.14EA 725 SEK
L06623.14 L06623.22
2-Chloro-5-nitrobenzotrifluoride ≥98%
2-Chloro-5-nitrobenzotrifluoride
Purity: 98% CAS No.: 777-37-7 Molecular Formula: C7H3ClF3NO2 Formula Weight: 225.55 Flash Point: 98°(208°F) Refractive Index: 1.5080 Melting Point: ca 22° Boiling Point: 232° Beilstein Reference: 2216169 MDL No.: MFCD00007296
Formel: C₇H₃ClF₃NO₂
MW: 225,55 g/mol
Kokpunkt: 232 °C
Smältpunkt: ∼22 °C
Densitet: 1,547
Flampunkt: 98 °C (208 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007296
CAS nummer: 777-37-7
EINECS: 212-287-1