4-Fluorobenzyl chloride 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Fluorobenzyl chloride, p-Fluorobenzyl chloride, 1-Chloromethyl-4-fluorobenzene, α-Chloro-4-fluorotoluene, 1-(Chloromethyl)-4-fluorobenzene
119430250 119431000
ACRO119430250EA 805 SEK
ACRO119430250 ACRO119431000
4-Fluorobenzyl chloride 98%
4-Fluorobenzyl chloride
Formel: C₇H₆ClF
Kokpunkt: 181,2 °C (1013 hPa)
Smältpunkt: –18 °C
Densitet: 1,207 g/cm³
Flampunkt: 72 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000913
CAS nummer: 352-11-4
EINECS: 206-516-4
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5120 to 1.5140 (20°C, 589 nm)
GC ≥97.5 %