3-Nitro-p-toluic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Methyl-3-nitrobenzoic acid
A11650.22 A11650.36
A11650.22EA 1375 SEK
A11650.22 A11650.36
3-Nitro-p-toluic acid 99%
3-Nitro-p-toluic acid
Purity: 99% CAS No.: 96-98-0 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 188-190° Beilstein Reference: 1874411
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Kokpunkt: 360 °C (1013 hPa)
Smältpunkt: 188…190 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007174
CAS nummer: 96-98-0
EINECS: 202-549-3