6-Nitro-m-toluic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Nitro-5-methylbenzoic acid, 5-Methyl-2-nitrobenzoic acid
L06594.06 L06594.14
L06594.06EA 1275 SEK
L06594.06 L06594.14
6-Nitro-m-toluic acid ≥98%
6-Nitro-m-toluic acid
Purity: 98+% CAS No.: 3113-72-2 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 133-137° Beilstein Reference: 1959031
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Kokpunkt: 364 °C (1013 hPa)
Smältpunkt: 133…137 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007368
CAS nummer: 3113-72-2
EINECS: 221-481-5