5-Methylanthranil acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Amino-5-methylbenzoic acid, 6-Amino-m-toluic acid
L01513.06 L01513.14
L01513.06EA 2650 SEK
L01513.06 L01513.14
5-Methylanthranil acid 97%
5-Methylanthranil acid
Purity: 97% CAS No.: 2941-78-8 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Melting Point: ca 175° dec. Beilstein Reference: 1101527
Formel: C₈H₉NO₂
MW: 151,16 g/mol
Kokpunkt: 317 °C (1013 hPa)
Smältpunkt: ∼ 175 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007909
CAS nummer: 2941-78-8
EINECS: 220-932-3