4-Pentylbenzoic acid
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
4-Amylbenzoic acid, 4-n-Pentylbenzoic acid, 4-Pentylbenzoic acid
BTB09863DA
BTB09863EA
MAYBBTB09863DAEA
0
SEK
MAYBBTB09863DA
MAYBBTB09863EA
4-Pentylbenzoic acid
4-Pentylbenzoic acid
Formel:
C₁₂H₁₆O₂ MW: 192.26 g/mol Kokpunkt: 260.7 Smältpunkt: 85…88 °C Densitet: ∼0.992 g/cm³ (20 °C) |
MDL Number:
MFCD00002572 CAS nummer: 26311-45-5 EINECS: 247-607-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...