p-Toluic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Methylbenzoic acid, 4-Toluic acid
139060010 139061000 139062500
ACRO139060010EA 2300 SEK
ACRO139060010 ACRO139061000 ACRO139062500
p-Toluic acid 98%
p-Toluic acid
Formel: C₈H₈O₂
MW: 136,15 g/mol
Kokpunkt: 274…275 °C (760 mmHg)
Smältpunkt: 179…182 °C
Densitet: 1,06 g/cm³ (20 °C)
Flampunkt: 180 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002565
CAS nummer: 99-94-5
EINECS: 202-803-3
Merck Index: 12,09673
Order Now
Specifikation testresultat
Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 179 to 182 °C
Titration with NaOH ≥97.5 %
Elemental Analysis 41.2 to 46.4 % (O)