o-Toluic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Methylbenzoic acid
A13856.0B A13856.30
A13856.0BEA 1600 SEK
A13856.0B A13856.30
o-Toluic acid ≥98%
o-Toluic acid
Purity: 98+% CAS No.: 118-90-1 Molecular Formula: C8H8O2 Formula Weight: 136.15 Flash Point: 148°(298°F) Odour: Aromatic Boiling Point: 258-259° Beilstein Reference: 1072103 Merck Reference: 14,9535 MDL No.: MFCD00002477
Formel: C₈H₈O₂
MW: 136,15 g/mol
Kokpunkt: 258…259 °C
Smältpunkt: 103…106 °C
Densitet: 1,060
Flampunkt: 148 °C (298 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002477
CAS nummer: 118-90-1
EINECS: 204-284-9
Merck Index: 14,9535