o-Toluic acid 98+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Methylbenzoic acid
139040010 139040050 139041000
ACRO139040010EA 1825 SEK
ACRO139040010 ACRO139040050 ACRO139041000
o-Toluic acid 98+%
o-Toluic acid
Formel: C₈H₈O₂
MW: 136.15 g/mol
Kokpunkt: 259
Smältpunkt: 103…105 °C
Flampunkt: 148
Storage Temperature: Omgivande
MDL Number: MFCD00002477
CAS nummer: 118-90-1
EINECS: 204-284-9
Merck Index: 12,09673
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 102 to 106 °C
Titration with NaOH ≥98.0 %