4-Methoxyphenylacetone 98+%
Leverantör: Thermo Scientific
Synonyms: 1-(p-Methoxyphenyl)-2-propanone, 1-(4-Methoxyphenyl)-2-propanone
189800500 189800050 189802500
ACRO189800500EA 1075 SEK
ACRO189800500 ACRO189800050 ACRO189802500
4-Methoxyphenylacetone 98+%
4-Methoxyphenylacetone
Formel: C₁₀H₁₂O₂
MW: 164,2 g/mol
Kokpunkt: 266…268 °C (760 mmHg)
Smältpunkt: -15 °C
Densitet: 1,067 g/cm³
Flampunkt: 135 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008773
CAS nummer: 122-84-9
EINECS: 204-578-7
Order Now
Specifikation testresultat
Appearance Clear light yellow to yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5300 (20°C, 589 nm)
GC ≥98.0 %