Nonanophenone ≥97%
Leverantör: Thermo Fisher Scientific
A12238.09 A12238.18
A12238.09EA 1125 SEK
A12238.09 A12238.18
Nonanophenone ≥97%
Nonanophenone
Purity: 97% CAS No.: 6008-36-2 Molecular Formula: C15H22O Formula Weight: 218.34 Flash Point: 110°(230°F) Refractive Index: 1.5060 Melting Point: 14° Boiling Point: 139-140°/2mm Beilstein Reference: 2047454 MDL No.: MFCD00048965
Formel: C₆H₅C(O)(CH₂)₇CH₃
Kokpunkt: 139…140 °C (2 mmHg)
Smältpunkt: 14 °C
Densitet: 0,940
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00048965
CAS nummer: 6008-36-2
EINECS: 227-861-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=954329dfa1dfb3703f537ff434c29d3d07d6dc94&vid=8404e060bb250a6a8e0cec57e16cf8c5c1d53d5a&prid=web&clid=SAPDAM