Butyrophenone 99%
Leverantör: Thermo Scientific
Synonyms: 1-Phenylbutan-1-one, Phenyl propyl ketone, 1-Phenyl-1-butanone
154411000 154415000
ACRO154411000EA 1175 SEK
ACRO154411000 ACRO154415000
Butyrophenone 99%
Butyrophenone
Formel: C₆H₅C(O)(CH₂)₂CH₃
MW: 148,2 g/mol
Kokpunkt: 228…230 °C (760 mmHg)
Smältpunkt: 11…13 °C
Densitet: 0,988 g/cm³
Flampunkt: 88 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009397
CAS nummer: 495-40-9
EINECS: 207-799-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74d21a84719c71ec28f51844913f7376027ac053&vid=ee1190f31dd1e4417139334f6b18ee0a34ddd5f8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5185 to 1.5205 (20°C, 589 nm)
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure