4-Pentylbenzonitrile

Leverantör: Thermo Scientific

BTB12834DA BTB12834EA
MAYBBTB12834DAEA 0 SEK
MAYBBTB12834DA MAYBBTB12834EA
4-Pentylbenzonitrile
4-Pentylbenzonitrile
Formel: C₁₂H₁₅N
MW: 173.26 g/mol
CAS nummer: 10270-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR