4-Bromo-3-nitrotoluene ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-Bromo-4-methyl-2-nitrobenzene
A10718.14 A10718.22
A10718.14EA 1675 SEK
A10718.14 A10718.22
4-Bromo-3-nitrotoluene ≥98%
4-Bromo-3-nitrotoluene
Purity: 98+% CAS No.: 5326-34-1 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Flash Point: 110°(230°F) Melting Point: 29-32° Boiling Point: 96°/1mm Beilstein Reference: 1945925 MDL No.: MFCD00024180
Formel: C₇H₆BrNO₂
MW: 216.03 g/mol
Kokpunkt: 96 °C (1 mmHg)
Smältpunkt: 29…32 °C
Densitet: 1.578
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00024180
CAS nummer: 5326-34-1
EINECS: 226-203-6