4-Bromo-α,α,α-trifluorotoluene 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene
166670050 166670250
ACRO166670050EA 895 SEK
ACRO166670050 ACRO166670250
4-Bromo-α,α,α-trifluorotoluene 99%
4-Bromo-α,α,α-trifluorotoluene
Formel: C₇H₄BrF₃
MW: 225,01 g/mol
Kokpunkt: 154…155 °C (760 mmHg)
Densitet: 1,6 g/cm³
Flampunkt: 48 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000398
CAS nummer: 402-43-7
EINECS: 206-943-6
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4715 to 1.4745 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥98.5 %