4-Bromo-2-fluorotoluene ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Bromo-3-fluoro-4-methylbenzene
A15891.06 A15891.14 A15891.22
A15891.06EA 655 SEK
A15891.06 A15891.14 A15891.22
4-Bromo-2-fluorotoluene ≥99%
4-Bromo-2-fluorotoluene
Purity: 99% CAS No.: 51436-99-8 Molecular Formula: C7H6BrF Formula Weight: 189.03 Flash Point: 73°(163°F) Refractive Index: 1.5290 Boiling Point: 66-68°/8mm Beilstein Reference: 1859028 MDL No.: MFCD00013551
Formel: C₇H₆BrF
Kokpunkt: 66…68 °C (8 mmHg)
Densitet: 1,492
Flampunkt: 73 °C (163 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00013551
CAS nummer: 51436-99-8
EINECS: 257-201-3