α-Bromo-4-fluorotoulene 97%
Leverantör: Thermo Scientific
Fara
Synonyms: α-Bromo-p-fluorotoulene, 4-Fluorobenzyl bromide, p-Fluorobenzyl bromide
197970050 197970250
ACRO197970050EA 695 SEK
ACRO197970050 ACRO197970250
α-Bromo-4-fluorotoulene 97%
α-Bromo-4-fluorotoulene
Formel: C₇H₆BrF
MW: 189,03 g/mol
Kokpunkt: 85 °C (15 mmHg)
Densitet: 1,517 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000359
CAS nummer: 459-46-1
EINECS: 207-291-5
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5464 to 1.5484 (20°C, 589 nm)
GC ≥96.0 %