α-Bromo-2,5-difluorotoluene ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-(Bromomethyl)-1,4-difluorobenzene, 2,5-Difluorobenzyl bromide
B20752.09 B20752.18 B20752.88
B20752.09EA 2175 SEK
B20752.09 B20752.18 B20752.88
α-Bromo-2,5-difluorotoluene ≥98%
α-Bromo-2,5-difluorotoluene
Purity: 98% CAS No.: 85117-99-3 Molecular Formula: C7H5BrF2 Formula Weight: 207.02 Refractive Index: 1.5260 Beilstein Reference: 7089248 UN No.: 3265 MDL No.: MFCD00009897
Formel: C₇H₅BrF₂
MW: 207.02 g/mol
Kokpunkt: 186
Smältpunkt: 8
Densitet: 1.609
Storage Temperature: Omgivande
MDL Number: MFCD00009897
CAS nummer: 85117-99-3
EINECS: 285-651-0
UN: 3265
ADR: 8 III