2,4-Difluorobenzyl bromide ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-(Bromomethyl)-2,4-difluorobenzene, α-Bromo-2,4-difluorotoluene
A15213.06 A15213.14
A15213.06EA 1325 SEK
A15213.06 A15213.14
2,4-Difluorobenzyl bromide ≥98%
2,4-Difluorobenzyl bromide
Purity: 98+% CAS No.: 23915-07-3 Molecular Formula: C7H5BrF2 Formula Weight: 207.02 Flash Point: 85°(185°F) Refractive Index: 1.5250 Boiling Point: 42-43°/1mm Beilstein Reference: 4177539 UN No.: 1760 MDL No.: MFCD00011649
Formel: C₇H₅BrF₂
MW: 207.02 g/mol
Kokpunkt: 42…43 °C (1 mmHg)
Densitet: 1.613
Flampunkt: 85 °C (185 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00011649
CAS nummer: 23915-07-3
EINECS: 245-938-3
UN: 1760
ADR: 8 III