α-Bromo-2-chlorotoulene 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide
278930250 278930050
ACRO278930250EA 2350 SEK
ACRO278930250 ACRO278930050
α-Bromo-2-chlorotoulene 97%
α-Bromo-2-chlorotoulene
Formel: C₇H₆BrCl
Kokpunkt: 103…104 °C (10 mmHg)
Densitet: 1,586 g/cm³
Flampunkt: 108 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000566
CAS nummer: 611-17-6
EINECS: 210-257-2
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5910 to 1.5930 (20°C, 589 nm)
GC ≥97.5 %