α,3-Dibromotoluene 95%
Leverantör: Thermo Scientific
Fara
Synonyms: 3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene
181780050 181780250
ACRO181780050EA 665 SEK
ACRO181780050 ACRO181780250
α,3-Dibromotoluene 95%
α,3-Dibromotoluene
Formel: C₇H₆Br₂
MW: 249,93 g/mol
Kokpunkt: 130 °C (12 mmHg)
Smältpunkt: 37…42 °C
Densitet: 1,56 g/cm³ (20 °C)
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000176
CAS nummer: 823-78-9
EINECS: 212-519-1
UN: 3261
ADR: 8,II
Order Now
Specifikation testresultat
Appearance White Adhering crystalline powder or crystals
Infrared spectrum Conforms
Melting point 37 to 42 °C
GC ≥94 %