4-Bromophenylboronic acid neopentyl glycol ester ≥98%
Leverantör: Thermo Fisher Scientific
Synonyms: 2-(4-Bromophenyl)-5,5-dimethyl-1,3,2-dioxaborinane, 4-Bromobenzeneboronic acid neopentyl glycol ester
L17455.03 L17455.06
L17455.03EA 645 SEK
L17455.03 L17455.06
4-Bromophenylboronic acid neopentyl glycol ester ≥98%
4-Bromophenylboronic acid neopentyl glycol ester
Purity: 98+% CAS No.: 183677-71-6 Molecular Formula: C11H14BBrO2 Formula Weight: 268.95 Melting Point: 109-111° MDL No.: MFCD01318105
Formel: C₁₁H₁₄BBrO₂
MW: 268.94 g/mol
Smältpunkt: 109…111 °C
Storage Temperature: Omgivande
MDL Number: MFCD01318105
CAS nummer: 183677-71-6