4-Cyanophenylboronic acid pinacol ester ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-Cyanobenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyanophenylboronic acid, pinacol ester
H28208.06 H28208.03
H28208.06EA 4675 SEK
H28208.06 H28208.03
4-Cyanophenylboronic acid pinacol ester ≥97%
4-Cyanophenylboronic acid pinacol ester
Purity: 97% CAS No.: 171364-82-2 Molecular Formula: C13H16BNO2 Formula Weight: 229.08 MDL No.: MFCD03093897
Formel: C₁₃H₁₆BNO₂
MW: 229,09 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03093897
CAS nummer: 171364-82-2