Phenethyl bromide ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: (2-Bromoethyl)benzene, 2-Phenylethyl bromide
A12528.22 A12528.36
A12528.22EA 725 SEK
A12528.22 A12528.36
Phenethyl bromide ≥98%
Phenethyl bromide
Purity: 98% CAS No.: 103-63-9 Molecular Formula: C8H9Br Formula Weight: 185.06 Flash Point: 89°(192°F) Refractive Index: 1.5560 Melting Point: -56° Boiling Point: 220-221° Beilstein Reference: 507487 MDL No.: MFCD00000240
Formel: BrCH₂CH₂C₆H₅
MW: 185,06 g/mol
Kokpunkt: 220…221 °C
Smältpunkt: –56 °C
Densitet: 1,355
Flampunkt: 89 °C (192 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000240
CAS nummer: 103-63-9
EINECS: 203-130-8