1-Phenyl-1-propyne 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Propynylbenzene, Prop-1-yn-1-ylbenzene
171320050 171320250
ACRO171320050EA 1350 SEK
ACRO171320050 ACRO171320250
1-Phenyl-1-propyne 99%
1-Phenyl-1-propyne
Formel: C₆H₅CCCH₃
MW: 116,16 g/mol
Kokpunkt: 185 °C (760 mmHg)
Densitet: 0,92 g/cm³
Flampunkt: 62 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009272
CAS nummer: 673-32-5
EINECS: 211-607-7
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5650 (20°C, 589 nm)
GC ≥98.5 %