1-Butynylbenzene ≥98%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
1-Phenyl-1-butyne
A11332.06
A11332.09
A11332.14
A11332.06EA
965
SEK
A11332.06
A11332.09
A11332.14
1-Butynylbenzene ≥98%
1-Butynylbenzene
Purity: 98%
CAS No.: 622-76-4
Molecular Formula: C10H10
Formula Weight: 130.19
Flash Point: 68°(154°F)
Refractive Index: 1.5520
Boiling Point: 201-203°
Beilstein Reference: 1903397
MDL No.: MFCD00039945
Formel:
C₆H₅CCCH₂CH₃ MW: 130,19 g/mol Kokpunkt: 201…203 °C Densitet: 0,918 Flampunkt: 68 °C (154 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00039945 CAS nummer: 622-76-4 EINECS: 210-752-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...