Triphenylethylene ≥98%
Leverantör: Thermo Scientific
Synonyms: 1,1,2-Triphenylethylene, 1,1,2-Triphenylethene
A13244.09 A13244.18
A13244.09EA 1125 SEK
A13244.09 A13244.18
Triphenylethylene ≥98%
Triphenylethylene
Purity: 98+% CAS No.: 58-72-0 Molecular Formula: C20H16 Formula Weight: 256.35 Melting Point: 67-71° Boiling Point: 219-221°/14mm MDL No.: MFCD00004765
Formel: (C₆H₅)₂CCHC₆H₅
MW: 256,35 g/mol
Kokpunkt: 219…221 °C (14 mmHg)
Smältpunkt: 67…71 °C
Densitet: 1,0373 g/cm³ (78.4 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004765
CAS nummer: 58-72-0
EINECS: 200-395-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f975e70dcf0b39f840f804277fd5a955f70445bd&vid=59d8884b3eb04ca490c5831fb69542a8eed463ad&prid=web&clid=SAPDAM