2-Fenylpropen ≥99% stabilised
Leverantör: Thermo Fisher Scientific
|
Varning
|
Synonyms:
Isopropenylbenzen
Stabilization:
Stabiliseras med 4-tert-Butylpyrokatekol
L03609.AE
L03609.AP
L03609.0F
L03609.AEEA
570
SEK
L03609.AE
L03609.AP
L03609.0F
2-Fenylpropen ≥99% stabilised
2-Fenylpropen
Stabilized with 10-20 ppm 4-tert-butylcatechol
Purity: 99%
CAS No.: 98-83-9
Molecular Formula: C9H10
Formula Weight: 118.18
Flash Point: 45°(113°F)
Refractive Index: 1.5380
Melting Point: -23°
Boiling Point: 166-169°
Beilstein Reference: 969405
UN No.: 2303
MDL No.: MFCD00008859
Formel:
C₉H₁₀ MW: 118.18 g/mol Kokpunkt: 166…169 °C Smältpunkt: –23 °C Densitet: 0.909 Flampunkt: 45 °C (113 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00008859 CAS nummer: 98-83-9 EINECS: 202-705-0 UN: 2303 ADR: 3 III |