1-Aminocyclopropanecarboxylic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Amino-1-cyclopropanecarboxylic acid
301422500 301420010
ACRO301422500EA 3100 SEK
ACRO301422500 ACRO301420010
1-Aminocyclopropanecarboxylic acid 99%
1-Aminocyclopropanecarboxylic acid
Formel: C₄H₇NO₂
MW: 101,11 g/mol
Smältpunkt: 257 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00009944
CAS nummer: 22059-21-8
Order Now
Specifikation testresultat
Appearance White to off-white Powder and/or granules
Infrared spectrum Conforms
Water ≤9 % (K.F.)
Titration with NaOH ≥98.5 % (On dry substance)