4'-Hexylbiphenyl-4-carboxylic acid ≥99%

Leverantör: Thermo Scientific
Varning

Synonyms: 4'-Hexyl-[1,1'-biphenyl]-4-carboxylic acid

B21900.03 B21900.06 B21900.14
B21900.03EA 695 SEK
B21900.03 B21900.06 B21900.14
4'-Hexylbiphenyl-4-carboxylic acid ≥99%
4'-Hexylbiphenyl-4-carboxylic acid

Purity: 99% CAS No.: 59662-48-5 Molecular Formula: C19H22O2 Formula Weight: 282.38 Melting Point: 222-224° MDL No.: MFCD00143232

Formel: C₁₉H₂₂O₂
MW: 282,38 g/mol
Smältpunkt: 222…224 °C
Storage Temperature: Omgivande
MDL Number: MFCD00143232
CAS nummer: 59662-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR