(±)-1-Phenyl-2-propyn-1-ol 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Phenyl-2-propyn-1-ol
L09549.03 L09549.06
L09549.03EA 735 SEK
L09549.03 L09549.06
(±)-1-Phenyl-2-propyn-1-ol 98%
(±)-1-Phenyl-2-propyn-1-ol
Purity: 98% CAS No.: 4187-87-5 Molecular Formula: C9H8O Formula Weight: 132.16 Flash Point: 99°(210°F) Refractive Index: 1.5500 Melting Point: 26-30° Boiling Point: 231-233° Beilstein Reference: 742365 MDL No.: MFCD00021860
Formel: C₉H₈O
MW: 132,16 g/mol
Kokpunkt: 231…233 °C
Smältpunkt: 26…30 °C
Densitet: 1,087
Flampunkt: 99 °C (210 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00021860
CAS nummer: 4187-87-5
EINECS: 224-064-6