2-Aminophenylboronic acid pinacol ester 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester
L19951.03 L19951.06
L19951.03EA 2175 SEK
L19951.03 L19951.06
2-Aminophenylboronic acid pinacol ester 97%
2-Aminophenylboronic acid pinacol ester
Purity: 97% CAS No.: 191171-55-8 Molecular Formula: C12H18BNO2 Formula Weight: 219.09 Melting Point: 65-70° Beilstein Reference: 8548809
Formel: C₁₂H₁₈BNO₂
MW: 219,09 g/mol
Smältpunkt: 65…70 °C
Storage Temperature: Omgivande
MDL Number: MFCD02179448
CAS nummer: 191171-55-8