1-Butylboronic acid, Supelco®

Leverantör: Merck

Synonyms: Butylboronic acid, n-Butylboronic acid

M-Clarity™ quality level = MQ100

19667-5G
19667-5GEA 3120 SEK
19667-5G
1-Butylboronic acid, Supelco®
1-Butylboronic acid
Formel: C₄H₁₁BO₂
MW: 101.94 g/mol
MDL Number: MFCD00002106
CAS nummer: 4426-47-5
EINECS: 224-607-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR