4-Amino-2-(trifluoromethyl)benzonitrile 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-Cyano-3-trifluoromethylaniline
L12346.03 L12346.MD L12346.06
L12346.03EA 990 SEK
L12346.03 L12346.MD L12346.06
4-Amino-2-(trifluoromethyl)benzonitrile 97%
4-Amino-2-(trifluoromethyl)benzonitrile
Purity: 97% CAS No.: 654-70-6 Molecular Formula: C8H5F3N2 Formula Weight: 186.14 Melting Point: 136-139° Beilstein Reference: 3278074 UN No.: 3439 MDL No.: MFCD00042155
Formel: C₈H₅F₃N₂
MW: 186,14 g/mol
Smältpunkt: 136…139 °C
Storage Temperature: Omgivande
MDL Number: MFCD00042155
CAS nummer: 654-70-6
UN: 3439
ADR: 6.1,III