3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide
L11939.03 L11939.06
L11939.03EA 1125 SEK
L11939.03 L11939.06
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98%
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Purity: 98% CAS No.: 298-93-1 Molecular Formula: C18H16BrN5S Formula Weight: 414.33 Odour: Odourless Melting Point: ca 195° dec. Beilstein Reference: 3825277 MDL No.: MFCD00011964
Formel: C₁₈H₁₆N₅BrS
MW: 414,33 g/mol
Smältpunkt: ∼ 195 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00011964
CAS nummer: 298-93-1
EINECS: 206-069-5