2,3-Dihydrofuran 98+%
Leverantör: Thermo Fisher Scientific
Fara
190441000 190445000
ACRO190441000EA 1500 SEK
ACRO190441000 ACRO190445000
2,3-Dihydrofuran 98+%
2,3-Dihydrofuran
Formel: C₄H₆O
MW: 70.09 g/mol
Densitet: 0.927 g/cm³
Flampunkt: –24 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00003205
CAS nummer: 1191-99-7
EINECS: 214-747-7
UN: 1993
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4250 (20°C, 589 nm)
Water ≤0.3 %
GC ≥98.0 %
Tetrahydrofuran ≤1.0 % (GC)