DL-α-Tocopherol acetate 97%
Leverantör: Thermo Fisher Scientific
Synonyms: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
A14505.14 A14505.22 A14505.36
A14505.14EA 1075 SEK
A14505.14 A14505.22 A14505.36
DL-α-Tocopherol acetate 97%
DL-α-Tocopherol acetate
Purity: 97% CAS No.: 7695-91-2 Molecular Formula: C31H52O3 Formula Weight: 472.76 Storage Temperature: 2°C to 8°C Flash Point: 210°(410°F) Refractive Index: 1.4960 Melting Point: -28° Boiling Point: 300° Beilstein Reference: 97512 Merck Reference: 14,9495 MDL No.: MFCD00072042
Formel: C₃₁H₅₂O₃
Kokpunkt: >300 °C
Smältpunkt: –28 °C
Densitet: 0,960
Flampunkt: 210 °C (410 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00072042
CAS nummer: 7695-91-2
EINECS: 231-710-0
Merck Index: 14,9495