Dimethyl bromomalonate 90% tech.
Leverantör: Thermo Fisher Scientific
Fara
305190050 305190250
ACRO305190050EA 470 SEK
ACRO305190050 ACRO305190250
Dimethyl bromomalonate 90% tech.
Dimethyl bromomalonate
Formel: C₅H₇BrO₄
Kokpunkt: 105…108 °C (11 mmHg)
Densitet: 1,6 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00025865
CAS nummer: 868-26-8
EINECS: 212-775-4
UN: 1760
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4660 (20°C, 589 nm)
GC ≥88 %