1-Bromo-2-methoxyethane 96% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Bromoethyl methyl ether
Stabilization: Stabilised with sodium carbonate (Na₂CO₃)
A15370.06 A15370.14 A15370.22 A15370.36
A15370.06EA 1125 SEK
A15370.06 A15370.14 A15370.22 A15370.36
1-Bromo-2-methoxyethane 96% stabilised
1-Bromo-2-methoxyethane
Stabilized with sodium carbonate
Purity: 96% CAS No.: 6482-24-2 Molecular Formula: BrCH2CH2OCH3 Formula Weight: 139.00 Storage Temperature: 2°C to 8°C Flash Point: 28°(82°F) Refractive Index: 1.4480 Odour: Recognizable Boiling Point: 109-110° UN No.: 3271 MDL No.: MFCD00000236
Formel: BrH₂CCH₂OCH₃
MW: 138.99 g/mol
Kokpunkt: 109…110 °C
Densitet: 1.478
Flampunkt: 28 °C (82 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00000236
CAS nummer: 6482-24-2
EINECS: 229-339-4
UN: 3271
ADR: 3 III