1-Boc-piperazine 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate
367280050 367280250
ACRO367280050EA 2725 SEK
ACRO367280050 ACRO367280250
1-Boc-piperazine 97%
1-Boc-piperazine
Formel: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Smältpunkt: 43…47 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00075265
CAS nummer: 57260-71-6
Order Now
Specifikation testresultat
Appearance White to light yellow Low melting crystalline mass
Infrared spectrum Conforms
GC ≥96.0 %