3,4,5-Trimethoxyphenol ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Antiarol
L01527.06 L01527.14
L01527.06EA 1850 SEK
L01527.06 L01527.14
3,4,5-Trimethoxyphenol ≥97%
3,4,5-Trimethoxyphenol
Purity: 97% CAS No.: 642-71-7 Molecular Formula: C9H12O4 Formula Weight: 184.19 Melting Point: 143-148° Beilstein Reference: 1958573
Formel: C₉H₁₂O₄
MW: 184,19 g/mol
Kokpunkt: 283 °C (1013 hPa)
Smältpunkt: 143…148 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008389
CAS nummer: 642-71-7
EINECS: 211-387-2