Geraniol 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-ol
410900250 410901000 410900050 410905000 410900025
ACRO410900250EA 900 SEK
ACRO410900250 ACRO410901000 ACRO410900050 ACRO410905000 ACRO410900025
Geraniol 99%
Geraniol
Formel: C₁₀H₁₈O
MW: 154.25 g/mol
Kokpunkt: 229…230 °C (757 mmHg)
Smältpunkt: –15 °C
Densitet: 0.878 g/cm³
Flampunkt: 76 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002917
CAS nummer: 106-24-1
EINECS: 203-377-1
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4780 (20°C, 589 nm)
GC ≥98.5 %