2,6-Dichlorobenzaldehyde oxime

Leverantör: Thermo Scientific

Synonyms: 2,6-Dichlorobenzaldoxime

SPB05820DA SPB05820EA
MAYBSPB05820DAEA 0 SEK
MAYBSPB05820DA MAYBSPB05820EA
2,6-Dichlorobenzaldehyde oxime
2,6-Dichlorobenzaldehyde oxime
Formel: C₇H₅Cl₂NO
MDL Number: MFCD00013938
CAS nummer: 25185-95-9
EINECS: 246-720-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR