2,6-Dichlorobenzaldehyde oxime ≥97%
Leverantör: Thermo Scientific
Synonyms: 2,6-Dichlorobenzaldoxime
A11351.06 A11351.14 A11351.22
A11351.06EA 655 SEK
A11351.06 A11351.14 A11351.22
2,6-Dichlorobenzaldehyde oxime ≥97%
2,6-Dichlorobenzaldehyde oxime
Purity: 97% CAS No.: 25185-95-9 Molecular Formula: C7H5Cl2NO Formula Weight: 190.03 Melting Point: 147-151° Beilstein Reference: 2046662
Formel: C₇H₅Cl₂NO
Smältpunkt: 147…151 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013938
CAS nummer: 25185-95-9
EINECS: 246-720-0