1,3,5-Trimethoxybenzene 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: O,O,O-1,3,5-Trimethylresorcinol
160550500 160550100 160552500
ACRO160550500EA 1350 SEK
ACRO160550500 ACRO160550100 ACRO160552500
1,3,5-Trimethoxybenzene 99%
1,3,5-Trimethoxybenzene
Formel: C₉H₁₂O₃
MW: 168.19 g/mol
Kokpunkt: 255 °C (760 mmHg)
Smältpunkt: 50…53 °C
Flampunkt: 85 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008385
CAS nummer: 621-23-8
EINECS: 210-673-4
Order Now
Specifikation testresultat
Appearance White to cream to yellow Powder or crystals or flakes and/or chunks
Infrared spectrum Conforms
Melting point 50 to 54 °C
GC ≥98.5 %