4-Methyl-2-(trifluoromethyl)benzonitrile 98%
Leverantör: Thermo Fisher Scientific
Fara
H31904.03 H31904.06
H31904.03EA 1500 SEK
H31904.03 H31904.06
4-Methyl-2-(trifluoromethyl)benzonitrile 98%
4-Methyl-2-(trifluoromethyl)benzonitrile
Purity: 98% CAS No.: 261952-05-0 Molecular Formula: C9H6F3N Formula Weight: 185.15 Melting Point: 22-28° UN No.: 3439 MDL No.: MFCD01631596
Formel: C₉H₆F₃N
MW: 185.15 g/mol
Smältpunkt: 22…28 °C
Storage Temperature: Omgivande
MDL Number: MFCD01631596
CAS nummer: 261952-05-0
UN: 3439
ADR: 6.1 III