1-Methyl-3-propylimidazolium iodide ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Methyl-3-n-propylimidazolium iodide, 1-Methyl-3-propyl-1H-imidazol-3-ium iodide
H27682.06 H27682.18
H27682.06EA 1625 SEK
H27682.06 H27682.18
1-Methyl-3-propylimidazolium iodide ≥98%
1-Methyl-3-propylimidazolium iodide
Purity: 98% CAS No.: 119171-18-5 Molecular Formula: C7H13IN2 Formula Weight: 252.09 Flash Point: 110°(230°F) Boiling Point: 301-304° MDL No.: MFCD08276385
Formel: C₇H₁₃IN₂
MW: 252,1 g/mol
Kokpunkt: 301…304 °C
Densitet: 1,54
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD08276385
CAS nummer: 119171-18-5