1-Bromo-2-(2-methoxyethoxy)ethane tech. 90% stabilised, Technical Grade
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Bromo-3,6-dioxaheptane, 2-(2-Methoxyethoxy)ethyl bromide, m-PEG2-Br
Stabilization: Stabilised with sodium carbonate (Na₂CO₃)
L05406.06 L05406.14
L05406.06EA 1300 SEK
L05406.06 L05406.14
1-Bromo-2-(2-methoxyethoxy)ethane tech. 90% stabilised, Technical Grade
1-Bromo-2-(2-methoxyethoxy)ethane
Formel: BrH₂CCH₂OCH₂CH₂OCH₃
Kokpunkt: 156 °C
Smältpunkt: –23 °C
Densitet: 1,347
Flampunkt: 73 °C (163 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00000238
CAS nummer: 54149-17-6
EINECS: 259-000-6